Learn More
JG-98, MedChemExpress
MedChemExpress JG-98, an allosteric heat shock protein 70 (Hsp70) inhibitor, which binds tightly to a conserved site on Hsp70 and disrupts the Hsp70-Bag3 interaction. JG-98 shows anti-cancer activities affecting both cancer cells and tumor-associated macrophages.

2598.37 DKK - 9154.76 DKK
Specifications
Chemical Name or Material | JG-98 |
---|---|
Physical Form | Solid |
CAS | 1456551-16-8 |
Color | Orange |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16451127
![]() |
Medchem Express
HY-117282 |
50 mg |
9154.76 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16417627
![]() |
Medchem Express
HY-117282 |
5 mg |
2598.37 DKK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16404878
![]() |
Medchem Express
HY-117282 |
10 mg |
3958.00 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
JG-98, an allosteric heat shock protein 70 (Hsp70) inhibitor, which binds tightly to a conserved site on Hsp70 and disrupts the Hsp70-Bag3 interaction. JG-98 shows anti-cancer activities affecting both cancer cells and tumor-associated macrophages.Specifications
JG-98 | |
1456551-16-8 | |
Research | |
Cancer-programmed cell death | |
CN1/C(SC2=CC(Cl)=CC=C12)=C3S/C(N(CC)C\3=O)=C\C4=[N+](C=CS4)CC5=CC=CC=C5.[Cl-] | |
534.54 | |
Research |
Solid | |
Orange | |
C24H21Cl2N3OS3 | |
DMSO : 5 mg/mL (9.35 mM; Need ultrasonic) | |
534.54 | |
99.75% |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.