Få mere at vide
Isochlorogenic Acid B (>85%), TRC
Artikelnummer. 30009568
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30009568 2.5 mg 2.5mg
30007500 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30009568 Leverandør Toronto Research Chemicals Leverandørnr. TRCI7897002.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14534-61-3
Molekylær formel C25 H24 O12
Molekylvægt (g/mol) 516.45
Synonym Cyclohexanecarboxylic acid, 3,4-bis[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,5-dihydroxy-, (1S,3R,4R,5R)-, Cinnamic acid, 3,4-dihydroxy-, 5-carboxy-3,5-dihydroxy-1,2-cyclohexylene ester, stereoisomer (8CI), Cyclohexanecarboxylic acid, 3,4-bis[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,5-dihydroxy-, [1S-(1α,3β,4α,5α)]-, (1S,3R,4R,5R)-3,4-Bis[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,5-dihydroxycyclohexanecarboxylic acid, 3,4-Dicaffeoylquinic acid, Isochlorogenic acid b, Quinic acid 3,4-di-O-caffeate
IUPAC navn (1R,3R,4R,5R)-3,4-bis[[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy]-1,5-dihydroxycyclohexane-1-carboxylic acid
SMIL O[C@@H]1C[C@@](O)(C[C@@H](OC(=O)\C=C\c2ccc(O)c(O)c2)[C@@H]1OC(=O)\C=C\c3ccc(O)c(O)c3)C(=O)O
Tekniske data
Kemisk navn eller materiale Isochlorogenic acid
Mængde 2.5 mg
InChI formel InChI=1S/C25H24O12/c26-15-5-1-13(9-17(15)28)3-7-21(31)36-20-12-25(35,24(33)34)11-19(30)23(20)37-22(32)8-4-14-2-6-16(27)18(29)10-14/h1-10,19-20,23,26-30,35H,11-12H2,(H,33,34)/b7-3+,8-4+/t19-,20-,23-,25-/m1/s1
Formel vægt 516.1268