Learn More
Iron(II) iodide, ultra dry, 99.99% (metals basis)
Description
Iron(II) iodide is used as a catalyst in organic reactions. It is also used as a compound in semiconductors and other high purity applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Gray |
Physical Form | Beads |
Quantity | 1 g |
Assay Percent Range | 99.99% (Metal basis) |
Solubility Information | Soluble in water,diethyl ether and ethanol. |
Formula Weight | 309.66 |
Odor | Odorless |
Sensitivity | Air and light sensitive; Hygroscopic |
Appearance | Gray beads |
Density | 5.32 g/cm3 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
M30K026 | Certificate of Analysis | 05/02/2024 | 013619.09,013619.03 |
J03G21 | Certificate of Analysis | 07/12/2021 | 013619.09,013619.03 |
D05Y010 | Certificate of Analysis | 07/12/2021 | 013619.09,013619.03 |
N08F065 | Certificate of Analysis | 07/12/2021 | 013619.09,013619.03 |
B22F34 | Certificate of Analysis | 07/12/2021 | 013619.09,013619.03 |
Safety and Handling


- Iron(II) iodide anhydrous
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 C
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.