Få mere at vide
Irinotecan, MedChemExpress
Artikelnummer. 16491806
Encompass_Preferred
Skift visning
Mængde:
50 mg
100 mg
200 mg
500 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16491806 500 mg
16477116 100 mg
16473117 50 mg
16421226 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16491806 Leverandør Medchem Express Leverandørnr. HY16562
MedChemExpress Irinotecan ((+)-Irinotecan) is a topoisomerase I inhibitor, preventing religation of the DNA strand by binding to topoisomerase I-DNA complex.
Irinotecan ((+)-Irinotecan) is a topoisomerase I inhibitor, preventing religation of the DNA strand by binding to topoisomerase I-DNA complex.
Tekniske data
Kemisk navn eller materiale Irinotecan
CAS 97682-44-5
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C33H38N4O6
Mængde 500 mg
Synonym (+)-Irinotecan CPT-11
Opløselighedsinformation DMSO : 25 mg/mL (42.61 mM; Need ultrasonic)
SMIL O=C(N1CCC(N2CCCCC2)CC1)OC3=CC=C4N=C5C(CN6C(C(COC([C@@]7(CC)O)=O)=C7C=C65)=O)=C(CC)C4=C3
Molekylvægt (g/mol) 586.68
Formel vægt 586.68
Procent renhed 95.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death