Få mere at vide
Iodophenol Blue, TRC
Artikelnummer. 30415229
Skift visning
Mængde:
25 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30415229 25 mg
30417900 500 mg
30413131 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30415229 Leverandør Toronto Research Chemicals Leverandørnr. TRCT29618525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4430-24-4
Molekylær formel C14 H14 F N . H Cl
Molekylvægt (g/mol) 251.727
Synonym 3H-2,1-Benzoxathiole, Phenol deriv., 4,4'-(1,1-Dioxido-3H-2,1-benzoxathiol-3-ylidene)bis[2,6-diiodophenol], NSC 36792, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-diiodo-phenol S,S-Dioxide
IUPAC navn 2-[4-(4-fluorophenyl)phenyl]ethanamine;hydrochloride
SMIL Cl.NCCc1ccc(cc1)c2ccc(F)cc2
Tekniske data
Kemisk navn eller materiale Iodophenol Blue
Mængde 25 mg
InChI formel InChI=1S/C14H14FN.ClH/c15-14-7-5-13(6-8-14)12-3-1-11(2-4-12)9-10-16;/h1-8H,9-10,16H2;1H
Formel vægt 251.088