Få mere at vide
Indole-3-pyruvic Acid-d5, TRC
Artikelnummer. 30134566
Skift visning
Mængde:
1 mg
2.5 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134566 5 mg
30132625 2.5 mg
17927939 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30134566 Leverandør Toronto Research Chemicals Leverandørnr. TRCI6271825MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 85163-57-1
Molekylær formel C11H4D5NO3
Molekylvægt (g/mol) 208.22
Synonym (Penta-Deuteroindolyl-3)-pyruvic Acid;
IUPAC navn 2-oxo-3-(2,4,5,6,7-pentadeuterio-1H-indol-3-yl)propanoic acid
SMIL [2H]c1[nH]c2c([2H])c([2H])c([2H])c([2H])c2c1CC(=O)C(=O)O
Tekniske data
Kemisk navn eller materiale Indole-3-pyruvic Acid-d5
Mængde 5 mg
InChI formel InChI=1S/C11H9NO3/c13-10(11(14)15)5-7-6-12-9-4-2-1-3-8(7)9/h1-4,6,12H,5H2,(H,14,15)/i1D,2D,3D,4D,6D
Formel vægt 208.0896