missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IL11 (Human) Recombinant Protein
Human IL11 full-length ORF ( NP_000632, 1 a.a. - 199 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003589-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MNCVCRLVLVVLSLWPDTAVAPGPPPGPPRVSPDPRAELDSTVLLTRSLLADTRQLAAQLRDKFPADGDHNLDSLPTLAMSAGALGALQLPGVLTRLRADLLSYLRHVQWLRRAGGSSLKTLEPELGTLQARLDRLLRRLQLLMSRLALPQPPPDPPAPPLAPPSSAWGGIRAALAILGGLHLTLDWAVRGLLLLKTRL
Specifications
NP_000632 | |
interleukin 11 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IL11 | |
GST |
3589 | |
Wheat germ expression system | |
25 μg | |
AGIF, IL-11 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |