missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IL11 (Human) Recombinant Protein
Human IL11 full-length ORF ( NP_000632, 1 a.a. - 199 a.a.) recombinant protein with GST-tag at N-terminal.
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_000632 |
---|---|
Gene ID (Entrez) | 3589 |
Name | interleukin 11 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16180075
|
Abnova™
H00003589-P01.10UG |
10 μg |
2665.00 DKK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16190075
|
Abnova™
H00003589-P01.25UG |
25 μg |
4040.00 DKK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MNCVCRLVLVVLSLWPDTAVAPGPPPGPPRVSPDPRAELDSTVLLTRSLLADTRQLAAQLRDKFPADGDHNLDSLPTLAMSAGALGALQLPGVLTRLRADLLSYLRHVQWLRRAGGSSLKTLEPELGTLQARLDRLLRRLQLLMSRLALPQPPPDPPAPPLAPPSSAWGGIRAALAILGGLHLTLDWAVRGLLLLKTRL
Specifications
NP_000632 | |
interleukin 11 | |
125% SDS-PAGE Stained with Coomassie Blue | |
AGIF, IL-11 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
3589 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IL11 | |
GST |