Få mere at vide
Idasanutlin, MedChemExpress
Artikelnummer. 16442817
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16442817 50 mg
16439287 5 mg
16436738 10 mg
16406876 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16442817 Leverandør Medchem Express Leverandørnr. HY15676
MedChemExpress Idasanutlin (RG7388) is a potent and selective MDM2 antagonist, inhibiting p53-MDM2 binding, with an IC50 of 6 nM.
Idasanutlin (RG7388) is a potent and selective MDM2 antagonist, inhibiting p53-MDM2 binding, with an IC50 of 6 nM.
Tekniske data
Kemisk navn eller materiale Idasanutlin
CAS 1229705-06-9
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C31H29Cl2F2N3O4
Mængde 50 mg
Synonym RG7388
Opløselighedsinformation DMSO : ≥ 45 mg/mL (73.00 mM)
SMIL O=C(O)C1=CC(OC)=C(NC([C@H]2[C@H](C3=C(F)C(Cl)=CC=C3)[C@](C4=CC=C(Cl)C=C4F)(C#N)[C@H](CC(C)(C)C)N2)=O)C=C1
Molekylvægt (g/mol) 616.48
Formel vægt 616.48
Procent renhed 97.15%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death