Få mere at vide
Hyodeoxycholic acid, MedChemExpress
Artikelnummer. 16449016
Encompass_Preferred
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
16449016 100 mg
1 options
Artikelnummer. 16449016 Leverandør Medchem Express Leverandørnr. HYN0169
MedChemExpress Hyodeoxycholic acid is a secondary bile acid formed in the small intestine by the gut flora, and acts as a TGR5 (GPCR19) agonist, with an EC50 of 31.6 μM in CHO cells.
Hyodeoxycholic acid is a secondary bile acid formed in the small intestine by the gut flora, and acts as a TGR5 (GPCR19) agonist, with an EC50 of 31.6 μM in CHO cells.
Tekniske data
Kemisk navn eller materiale Hyodeoxycholic acid
CAS 83-49-8
Farve White
Renhedsgrad noter Research
Molekylær formel C24H40O4
Mængde 100 mg
Synonym HDCA
Opløselighedsinformation DMSO : ≥ 100 mg/mL (254.73 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMIL C[C@H](CCC(O)=O)[C@H]1CC[C@@]2([H])[C@]3([H])C[C@H](O)[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])CC[C@]12C
Molekylvægt (g/mol) 392.57
Formel vægt 392.57
Procent renhed 98.95%
Grad Research
Fysisk form Solid
Til brug med (applikation) Metabolism-sugar/lipid metabolism