Få mere at vide
Hydroxythiohomo Sildenafil, TRC
Artikelnummer. 30010016
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010016 1 mg
30008888 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30010016 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9631501MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 479073-82-0
Molekylær formel C23 H32 N6 O4 S2
Molekylvægt (g/mol) 520.67
Synonym 7H-Pyrazolo[4,3-d]pyrimidine-7-thione, 5-[2-ethoxy-5-[[4-(2-hydroxyethyl)-1-piperazinyl]sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-, 1-Piperazineethanol, 4-[[3-(4,7-dihydro-1-methyl-3-propyl-7-thioxo-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]- (9CI), 5-(2-ethoxy-5-(4-(2-hydroxyethyl)piperazin-1-ylsulfonyl)phenyl)-1-methyl-3-propyl-1H-pyrazolo[4,3-d]pyrimidine-7(6H)-thione, Hydroxythiohomosildenafil, Thiohydroxyhomosildenafil
IUPAC navn 5-[2-ethoxy-5-[4-(2-hydroxyethyl)piperazin-1-yl]sulfonylphenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidine-7-thione
SMIL CCCc1nn(C)c2C(=S)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(CCO)CC4
Tekniske data
Kemisk navn eller materiale Thiohydroxyhomosildenafil
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C23H32N6O4S2/c1-4-6-18-20-21(27(3)26-18)23(34)25-22(24-20)17-15-16(7-8-19(17)33-5-2)35(31,32)29-11-9-28(10-12-29)13-14-30/h7-8,15,30H,4-6,9-14H2,1-3H3,(H,24,25,34)
Formel vægt 520.1926
Procent renhed >95%