Få mere at vide
(Hydroxypropyl)methyl Cellulose Phthalate, TRC
Artikelnummer. 30138457
Skift visning
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138457 1 g
30133780 5 g
30005284 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138457 Leverandør Toronto Research Chemicals Leverandørnr. TRCH6260201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 9050-31-1
Molekylær formel C14H16N4
Molekylvægt (g/mol) 240.3
Synonym 2-Hydroxypropyl methyl cellulose phthalate, Cellulose methyl 2-hydroxypropyl ether hydrogen phthalate ester, Cellulose phthalate hydroxypropyl methyl ether, Glycidyl methacrylate-HPMCP copolymer, HP 5, HP 5 (cellulose derivative), HP 50;
SMIL CC(C)CN1C=NC2=C1C3=CC=CC=C3N=C2N
Tekniske data
Kemisk navn eller materiale Hypromellose phthalate
InChI formel InChI=1S/C14H16N4/c1-9(2)7-18-8-16-12-13(18)10-5-3-4-6-11(10)17-14(12)15/h3-6,8-9H,7H2,1-2H3,(H2,15,17)
Mængde 1 g