Få mere at vide
Hydroxychloroquine 3-Carbonitrile, TRC
Artikelnummer. 30179992
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179992 50 mg
30180301 10 mg
30180243 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179992 Leverandør Toronto Research Chemicals Leverandørnr. TRCH78393550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2054858-84-1
Molekylær formel C19H25ClN4O
Molekylvægt (g/mol) 360.88
Synonym 3-Cyano-Hydroxychloroquine, 7-Chloro-4-[[4-[ethyl(2-hydroxyethyl)amino]-1-methylbutyl]amino]-3-quinolinecarbonitrile
IUPAC navn 7-chloro-4-[5-[ethyl(2-hydroxyethyl)amino]pentan-2-ylamino]quinoline-3-carbonitrile
SMIL CCN(CCO)CCCC(C)Nc1c(cnc2cc(Cl)ccc12)C#N
Tekniske data
Kemisk navn eller materiale Hydroxychloroquine 3-Carbonitrile
Mængde 50 mg
InChI formel InChI=1S/C19H25ClN4O/c1-3-24(9-10-25)8-4-5-14(2)23-19-15(12-21)13-22-18-11-16(20)6-7-17(18)19/h6-7,11,13-14,25H,3-5,8-10H2,1-2H3,(H,22,23)
Formel vægt 360.1717