Learn More
Hamilton™ HVXM Large Body Modular Valve Positioner PTFE Valves
With a larger port internal diameter to allow increased fluid flows.
Brand:  Hamilton™ 36758
Additional Details : Weight : 0.06700kg
 View more versions of this product
Product Code. 11533274
  • 2085.00 DKK / Each
Estimated Shipment: 03-07-2024
to see stock.
Add to basket
Description
Description
  • Physically the largest valves
  • Even moderately viscous fluids flow freely
  • Exceptionally easy to service and modify
  • Maximizes flexibility and value
Specifications
Specifications
38°C
108.8 μL dead volume
Large body PTFE valve
3-5 model, 3 valves
1/4 in.-28
Aluminum Housing