Learn More
Hamilton™ HVXM Large Body Modular Valve Positioner PTFE Valves
With a larger port internal diameter to allow increased fluid flows.
2085.00 DKK - 2750.00 DKK
Specifications
Temperature (Metric) Max | 38°C |
---|---|
Volume (Metric) | 108.8 μL dead volume |
Thread Size | 1/4 in.-28 |
Description | Large body PTFE valve |
Material | Aluminum Housing |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
11533274
|
Hamilton™
36758 |
3-5 model, 3 valves |
2085.00 DKK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
11503364
|
Hamilton™
36759 |
4-5 model, 4 valves |
2750.00 DKK
Each |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
Description
- Physically the largest valves
- Even moderately viscous fluids flow freely
- Exceptionally easy to service and modify
- Maximizes flexibility and value
Specifications
38°C | |
1/4 in.-28 | |
Aluminum Housing |
108.8 μL dead volume | |
Large body PTFE valve |