Learn More
Hamilton™ HVXM Large Body Modular Valve Positioner PTFE Valves
With a larger port internal diameter to allow increased fluid flows.
Brand: Hamilton™ 36759
Additional Details : Weight : 0.07800kg
Description
- Physically the largest valves
- Even moderately viscous fluids flow freely
- Exceptionally easy to service and modify
- Maximizes flexibility and value
Specifications
38°C | |
108.8 μL dead volume | |
Large body PTFE valve |
4-5 model, 4 valves | |
1/4 in.-28 | |
Aluminum (Housing) |