Learn More
Abnova™ Human UPB1 Full-length ORF (AAI39841.1, 1 a.a. - 384 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051733-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that belongs to the CN hydrolase family. Beta-ureidopropionase catalyzes the last step in the pyrimidine degradation pathway. The pyrimidine bases uracil and thymine are degraded via the consecutive action of dihydropyrimidine dehydrogenase (DHPDH), dihydropyrimidinase (DHP) and beta-ureidopropionase (UP) to beta-alanine and beta-aminoisobutyric acid, respectively. UP deficiencies are associated with N-carbamyl-beta-amino aciduria and may lead to abnormalities in neurological activity. [provided by RefSeq]
Sequence: MAGAEWKSLEECLEKHLPLPDLQEVKRVLYGKELRKLDLPREAFEAASREDFELQGYAFEAAEEQLRRPRIVHVGLVQNRIPLPANAPVAEQVSALHRRIKAIVEVAAMCGVNIICFQEAWTMPFAFCTREKLPWTEFAESAEDGPTTRFCQKLAKNHDMVVVSPILERDSEHGDVLWNTAVVISNSGAVLGKTRKNHIPRVGDFNESTYYMEGNLGHPVFQTQFGRIAVNICYGRHHPLNWLMYSINGAEIIFNPSATIGALSESLWPIEARNAAIANHCFTCAINRVGTEHFPNEFTSGDGKKAHQDFGYFYGSSYVAAPDSSRTPGLSRSRDGLLVAKLDLNLCQQVNDVWNFKMTGRYEMYARELAEAVKSNYSPTIVKESpecifications
AAI39841.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68.64kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BUP1 | |
UPB1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
51733 | |
UPB1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAGAEWKSLEECLEKHLPLPDLQEVKRVLYGKELRKLDLPREAFEAASREDFELQGYAFEAAEEQLRRPRIVHVGLVQNRIPLPANAPVAEQVSALHRRIKAIVEVAAMCGVNIICFQEAWTMPFAFCTREKLPWTEFAESAEDGPTTRFCQKLAKNHDMVVVSPILERDSEHGDVLWNTAVVISNSGAVLGKTRKNHIPRVGDFNESTYYMEGNLGHPVFQTQFGRIAVNICYGRHHPLNWLMYSINGAEIIFNPSATIGALSESLWPIEARNAAIANHCFTCAINRVGTEHFPNEFTSGDGKKAHQDFGYFYGSSYVAAPDSSRTPGLSRSRDGLLVAKLDLNLCQQVNDVWNFKMTGRYEMYARELAEAVKSNYSPTIVKE | |
RUO | |
UPB1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |