missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human UPB1 Full-length ORF (AAI39841.1, 1 a.a. - 384 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | AAI39841.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51733 |
Molecular Weight (g/mol) | 68.64kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16180313
|
Abnova™
H00051733-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16170313
|
Abnova™
H00051733-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein that belongs to the CN hydrolase family. Beta-ureidopropionase catalyzes the last step in the pyrimidine degradation pathway. The pyrimidine bases uracil and thymine are degraded via the consecutive action of dihydropyrimidine dehydrogenase (DHPDH), dihydropyrimidinase (DHP) and beta-ureidopropionase (UP) to beta-alanine and beta-aminoisobutyric acid, respectively. UP deficiencies are associated with N-carbamyl-beta-amino aciduria and may lead to abnormalities in neurological activity. [provided by RefSeq]
Sequence: MAGAEWKSLEECLEKHLPLPDLQEVKRVLYGKELRKLDLPREAFEAASREDFELQGYAFEAAEEQLRRPRIVHVGLVQNRIPLPANAPVAEQVSALHRRIKAIVEVAAMCGVNIICFQEAWTMPFAFCTREKLPWTEFAESAEDGPTTRFCQKLAKNHDMVVVSPILERDSEHGDVLWNTAVVISNSGAVLGKTRKNHIPRVGDFNESTYYMEGNLGHPVFQTQFGRIAVNICYGRHHPLNWLMYSINGAEIIFNPSATIGALSESLWPIEARNAAIANHCFTCAINRVGTEHFPNEFTSGDGKKAHQDFGYFYGSSYVAAPDSSRTPGLSRSRDGLLVAKLDLNLCQQVNDVWNFKMTGRYEMYARELAEAVKSNYSPTIVKESpecifications
AAI39841.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68.64kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAGAEWKSLEECLEKHLPLPDLQEVKRVLYGKELRKLDLPREAFEAASREDFELQGYAFEAAEEQLRRPRIVHVGLVQNRIPLPANAPVAEQVSALHRRIKAIVEVAAMCGVNIICFQEAWTMPFAFCTREKLPWTEFAESAEDGPTTRFCQKLAKNHDMVVVSPILERDSEHGDVLWNTAVVISNSGAVLGKTRKNHIPRVGDFNESTYYMEGNLGHPVFQTQFGRIAVNICYGRHHPLNWLMYSINGAEIIFNPSATIGALSESLWPIEARNAAIANHCFTCAINRVGTEHFPNEFTSGDGKKAHQDFGYFYGSSYVAAPDSSRTPGLSRSRDGLLVAKLDLNLCQQVNDVWNFKMTGRYEMYARELAEAVKSNYSPTIVKE | |
RUO | |
UPB1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
51733 | |
UPB1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BUP1 | |
UPB1 | |
Recombinant | |
wheat germ expression system |