Learn More
Abnova™ Human SULT1B1 Full-length ORF (AAH10895, 1 a.a. - 296 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00027284-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. However, the total genomic length of this gene is greater than that of other SULT1 genes. [provided by RefSeq]
Sequence: MLSPKDILRKDLKLVHGYPMTCAFASNWEKIEQFHSRPDDIVIATYPKSGTTWVSEIIDMILNDGDIEKCKRGFITEKVPMLEMTPPGLRTSGIEQLEKNPSPRIVKTHLPTDLLPKSFWENNCKMIYLARNAKDVSVSYYHFDLMNNLQPFPGTWEEYLEKFLTGKVAYGSWFTHVKNWWKRKEEHPILFLYYEDMKENPKEEIKKIIRFLEKNLNDEILDRIIHHTSFEVMKDNPLVNYTHLPTTVMDHSKSPFMRKGTAGDWKNYFTVAQNEKFDAIYETEMSKTALQFRTEISpecifications
AAH10895 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
58.30kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC13356/ST1B2/SULT1B2 | |
SULT1B1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
27284 | |
SULT1B1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLSPKDILRKDLKLVHGYPMTCAFASNWEKIEQFHSRPDDIVIATYPKSGTTWVSEIIDMILNDGDIEKCKRGFITEKVPMLEMTPPGLRTSGIEQLEKNPSPRIVKTHLPTDLLPKSFWENNCKMIYLARNAKDVSVSYYHFDLMNNLQPFPGTWEEYLEKFLTGKVAYGSWFTHVKNWWKRKEEHPILFLYYEDMKENPKEEIKKIIRFLEKNLNDEILDRIIHHTSFEVMKDNPLVNYTHLPTTVMDHSKSPFMRKGTAGDWKNYFTVAQNEKFDAIYETEMSKTALQFRTEI | |
RUO | |
SULT1B1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |