missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SULT1B1 Full-length ORF (AAH10895, 1 a.a. - 296 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | AAH10895 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 27284 |
Molecular Weight (g/mol) | 58.30kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16178722
|
Abnova™
H00027284-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16168722
|
Abnova™
H00027284-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. However, the total genomic length of this gene is greater than that of other SULT1 genes. [provided by RefSeq]
Sequence: MLSPKDILRKDLKLVHGYPMTCAFASNWEKIEQFHSRPDDIVIATYPKSGTTWVSEIIDMILNDGDIEKCKRGFITEKVPMLEMTPPGLRTSGIEQLEKNPSPRIVKTHLPTDLLPKSFWENNCKMIYLARNAKDVSVSYYHFDLMNNLQPFPGTWEEYLEKFLTGKVAYGSWFTHVKNWWKRKEEHPILFLYYEDMKENPKEEIKKIIRFLEKNLNDEILDRIIHHTSFEVMKDNPLVNYTHLPTTVMDHSKSPFMRKGTAGDWKNYFTVAQNEKFDAIYETEMSKTALQFRTEISpecifications
AAH10895 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
58.30kDa | |
Glutathione Sepharose 4 Fast Flow | |
MLSPKDILRKDLKLVHGYPMTCAFASNWEKIEQFHSRPDDIVIATYPKSGTTWVSEIIDMILNDGDIEKCKRGFITEKVPMLEMTPPGLRTSGIEQLEKNPSPRIVKTHLPTDLLPKSFWENNCKMIYLARNAKDVSVSYYHFDLMNNLQPFPGTWEEYLEKFLTGKVAYGSWFTHVKNWWKRKEEHPILFLYYEDMKENPKEEIKKIIRFLEKNLNDEILDRIIHHTSFEVMKDNPLVNYTHLPTTVMDHSKSPFMRKGTAGDWKNYFTVAQNEKFDAIYETEMSKTALQFRTEI | |
RUO | |
SULT1B1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
27284 | |
SULT1B1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC13356/ST1B2/SULT1B2 | |
SULT1B1 | |
Recombinant | |
wheat germ expression system |