missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SEC14L4 Full-length ORF (CAK54647.1, 1 a.a. - 352 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00284904-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16139983

  • 4040.00 DKK / 25µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is highly similar to the protein encoded by the Saccharomyces cerevisiae SEC14 gene. The SEC14 protein is a phophatidylinositol transfer protein that is essential for biogenesis of Golgi-derived transport vesicles, and thus is required for the export of yeast secretory proteins from the Golgi complex. The specific function of this protein has not yet been determined. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: MLRRHMEFRKQQDLDNIVTWQPPEVIQLYDSGGLCGYDYEGCPVYFNIIGSLDPKGLLLSASKQDMIRKRIKVCELLLHECELQTQKLGRKIEMALMVFDMEGLSLKHLWKPAVEVYQQFFSILEANYPETLKNLIVIRAPKLFPVAFNLVKSFMSEETRRKIVILGDNWKQELTKFISPDQLPVEFGGTMTDPDGNPKCLTKINYGGEVPKSYYLCEQVRLQYEHTRSVGRGSSLQVENEILFPGCVLRWQFASDGGDIGFGVFLKTKMGEQQSAREMTEVLPSQRYNAHMVPEDGSLTCLQAGVYVLRFDNTYSRMHAKKLSYTVEVLLPDKASEETLQSLKAMRPSPTQ
Specifications

Specifications

CAK54647.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
65.12kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TAP3
SEC14L4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
284904
SEC14L4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLRRHMEFRKQQDLDNIVTWQPPEVIQLYDSGGLCGYDYEGCPVYFNIIGSLDPKGLLLSASKQDMIRKRIKVCELLLHECELQTQKLGRKIEMALMVFDMEGLSLKHLWKPAVEVYQQFFSILEANYPETLKNLIVIRAPKLFPVAFNLVKSFMSEETRRKIVILGDNWKQELTKFISPDQLPVEFGGTMTDPDGNPKCLTKINYGGEVPKSYYLCEQVRLQYEHTRSVGRGSSLQVENEILFPGCVLRWQFASDGGDIGFGVFLKTKMGEQQSAREMTEVLPSQRYNAHMVPEDGSLTCLQAGVYVLRFDNTYSRMHAKKLSYTVEVLLPDKASEETLQSLKAMRPSPTQ
RUO
SEC14L4
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers