missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SEC14L4 Full-length ORF (CAK54647.1, 1 a.a. - 352 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | CAK54647.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 284904 |
Molecular Weight (g/mol) | 65.12kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139983
|
Abnova™
H00284904-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16129983
|
Abnova™
H00284904-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is highly similar to the protein encoded by the Saccharomyces cerevisiae SEC14 gene. The SEC14 protein is a phophatidylinositol transfer protein that is essential for biogenesis of Golgi-derived transport vesicles, and thus is required for the export of yeast secretory proteins from the Golgi complex. The specific function of this protein has not yet been determined. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: MLRRHMEFRKQQDLDNIVTWQPPEVIQLYDSGGLCGYDYEGCPVYFNIIGSLDPKGLLLSASKQDMIRKRIKVCELLLHECELQTQKLGRKIEMALMVFDMEGLSLKHLWKPAVEVYQQFFSILEANYPETLKNLIVIRAPKLFPVAFNLVKSFMSEETRRKIVILGDNWKQELTKFISPDQLPVEFGGTMTDPDGNPKCLTKINYGGEVPKSYYLCEQVRLQYEHTRSVGRGSSLQVENEILFPGCVLRWQFASDGGDIGFGVFLKTKMGEQQSAREMTEVLPSQRYNAHMVPEDGSLTCLQAGVYVLRFDNTYSRMHAKKLSYTVEVLLPDKASEETLQSLKAMRPSPTQSpecifications
CAK54647.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.12kDa | |
Glutathione Sepharose 4 Fast Flow | |
MLRRHMEFRKQQDLDNIVTWQPPEVIQLYDSGGLCGYDYEGCPVYFNIIGSLDPKGLLLSASKQDMIRKRIKVCELLLHECELQTQKLGRKIEMALMVFDMEGLSLKHLWKPAVEVYQQFFSILEANYPETLKNLIVIRAPKLFPVAFNLVKSFMSEETRRKIVILGDNWKQELTKFISPDQLPVEFGGTMTDPDGNPKCLTKINYGGEVPKSYYLCEQVRLQYEHTRSVGRGSSLQVENEILFPGCVLRWQFASDGGDIGFGVFLKTKMGEQQSAREMTEVLPSQRYNAHMVPEDGSLTCLQAGVYVLRFDNTYSRMHAKKLSYTVEVLLPDKASEETLQSLKAMRPSPTQ | |
RUO | |
SEC14L4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
284904 | |
SEC14L4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TAP3 | |
SEC14L4 | |
Recombinant | |
wheat germ expression system |