Learn More
Abnova™ Human RPL41 Full-length ORF (-, 1 a.a. - 25 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
---|---|
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6171 |
Molecular Weight (g/mol) | 28.49kDa |
Name | RPL41 (Human) Recombinant Protein (P01) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16180312
|
Abnova™
H00006171-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16170312
|
Abnova™
H00006171-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein, which shares sequence similarity with the yeast ribosomal protein YL41, belongs to the L41E family of ribosomal proteins. It is located in the cytoplasm. The protein can interact with the beta subunit of protein kinase CKII and can stimulate the phosphorylation of DNA topoisomerase II-alpha by CKII. Two alternative splice variants have been identified, both encoding the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq]
Sequence: MRAKWRKKRMRRLKRKRRKMRQRSKSpecifications
Antibody Production, Protein Array, ELISA, Western Blot | |
6171 | |
RPL41 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RPL41 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
28.49kDa | |
Glutathione Sepharose 4 Fast Flow | |
MRAKWRKKRMRRLKRKRRKMRQRSK | |
RUO | |
RPL41 | |
Recombinant | |
wheat germ expression system |