Learn More
Abnova™ Human RPL41 Full-length ORF (-, 1 a.a. - 25 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006171-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein, which shares sequence similarity with the yeast ribosomal protein YL41, belongs to the L41E family of ribosomal proteins. It is located in the cytoplasm. The protein can interact with the beta subunit of protein kinase CKII and can stimulate the phosphorylation of DNA topoisomerase II-alpha by CKII. Two alternative splice variants have been identified, both encoding the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq]
Sequence: MRAKWRKKRMRRLKRKRRKMRQRSKSpecifications
Antibody Production, Protein Array, ELISA, Western Blot | |
6171 | |
RPL41 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRAKWRKKRMRRLKRKRRKMRQRSK | |
RUO | |
RPL41 | |
Recombinant | |
wheat germ expression system |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
28.49kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RPL41 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |