Learn More
Abnova™ Human PAX7 Partial ORF (NP_001128726.1, 351 a.a. - 434 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005081-Q02.10ug
Description
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 7 gene is unknown but speculated to involve tumor suppression since fusion of this gene with a forkhead domain family member has been associated with alveolar rhabdomyosarcoma. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: AYGARHSFSSYSDSFMNPAAPSNHMNPVSNGLSPQVMSILGNPSAVPPQPQADFSISPLHGGLDSATSISASCSQRADSIKPGDSpecifications
NP_001128726.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.87kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
AYGARHSFSSYSDSFMNPAAPSNHMNPVSNGLSPQVMSILGNPSAVPPQPQADFSISPLHGGLDSATSISASCSQRADSIKPGD | |
RUO | |
PAX7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5081 | |
PAX7 (Human) Recombinant Protein (Q02) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HUP1/PAX7B | |
PAX7 | |
Recombinant | |
wheat germ expression system |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.