missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human NRG1 Isoform GGF2 Antibody, R&D Systems™

Goat Polyclonal Antibody
1340.00 DKK - 3590.00 DKK
Tekniske data
| Antigen | Neuregulin-1 Isoform GGF2 |
|---|---|
| Koncentration | LYOPH |
| Fortynding | Western Blot 0.1 ug/mL |
| Applikationer | Western Blot |
| Klassifikation | Polyclonal |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
18707617
![]() |
R&D Systems
AF2015-SP |
25 μg |
1340.00 DKK
25µg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
18069161
![]() |
R&D Systems
AF2015 |
100 μg |
3590.00 DKK
100µg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
Neuregulin-1 Isoform GGF2 Polyclonal specifically detects Neuregulin-1 Isoform GGF2 in Human samples. It is validated for Western Blot.Tekniske data
| Neuregulin-1 Isoform GGF2 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| GGF2, ARIA, GGF, GGF2, Heregulin-1, HGL, HRG, HRG1, HRG1-alpha, HRG1-beta 1, HRGA, MST131, MSTP131, NDF, Neuregulin1, Neuregulin1 Isoform GGF2, NRG1, NRG1-IT2, SMDF | |
| NRG1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_039256 | |
| 3084 | |
| E. coli-derived recombinant human NRG1 Isoform GGF2 Gly51-Cys248 Accession # NP_039256 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human NRG1 Isoform GGF2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) NRG-1 alpha EGF domain, rhNRG1-beta 1 extracellular domain, rhNRG1-beta 1 EGF domain, rhSMDF, and recombinant mouse NRG3 is observed. |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel