missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human NRG1 Isoform GGF2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2015-SP
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Neuregulin-1 Isoform GGF2 Polyclonal specifically detects Neuregulin-1 Isoform GGF2 in Human samples. It is validated for Western Blot.
Tekniske data
| Neuregulin-1 Isoform GGF2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GGF2, ARIA, GGF, GGF2, Heregulin-1, HGL, HRG, HRG1, HRG1-alpha, HRG1-beta 1, HRGA, MST131, MSTP131, NDF, Neuregulin1, Neuregulin1 Isoform GGF2, NRG1, NRG1-IT2, SMDF | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3084 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| NP_039256 | |
| NRG1 | |
| E. coli-derived recombinant human NRG1 Isoform GGF2 Gly51-Cys248 Accession # NP_039256 | |
| 25 μg | |
| Primary | |
| Detects human NRG1 Isoform GGF2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) NRG-1 alpha EGF domain, rhNRG1-beta 1 extracellular domain, rhNRG1-beta 1 EGF domain, rhSMDF, and recombinant mouse NRG3 is observed. | |
| Human | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion