missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human LPAR5/LPA5/GPR92 Antibody, R&D Systems™

Mouse Monoclonal Antibody
1540.00 DKK - 3515.00 DKK
Tekniske data
| Antigen | LPAR5/LPA5/GPR92 |
|---|---|
| Klon | 459812 |
| Fortynding | Flow Cytometry 0.25 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL, CyTOF-ready |
| Applikationer | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF |
| Klassifikation | Monoclonal |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
18772207
![]() |
R&D Systems
MAB10297-SP |
25 μg |
1540.00 DKK
25µg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
18764657
![]() |
R&D Systems
MAB10297-100 |
100 μg |
3515.00 DKK
100µg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
LPAR5/LPA5/GPR92 Monoclonal specifically detects LPAR5/LPA5/GPR92 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.Tekniske data
| LPAR5/LPA5/GPR92 | |
| Flow Cytometry 0.25 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| G protein-coupled receptor 92, GPR92, GPR92G-protein coupled receptor 92, GPR93, GPR93G-protein coupled receptor 93, KPG_010, LPA receptor 5, LPA5, LPA5LPA-5, lysophosphatidic acid receptor 5 | |
| NS0 mouse myeloma cell line transfected with human LPAR5/LPA5/GPR92, Accession # Q9H1C0 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 459812 | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9H1C0 | |
| 57121 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel