missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human LPAR5/LPA5/GPR92 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10297-100
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
LPAR5/LPA5/GPR92 Monoclonal specifically detects LPAR5/LPA5/GPR92 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.
Tekniske data
| LPAR5/LPA5/GPR92 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| G protein-coupled receptor 92, GPR92, GPR92G-protein coupled receptor 92, GPR93, GPR93G-protein coupled receptor 93, KPG_010, LPA receptor 5, LPA5, LPA5LPA-5, lysophosphatidic acid receptor 5 | |
| NS0 mouse myeloma cell line transfected with human LPAR5/LPA5/GPR92, Accession # Q9H1C0 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
| 459812 | |
| Flow Cytometry 0.25 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Q9H1C0 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 57121 | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion