Learn More
Abnova™ Human LOC201181 Full-length ORF (NP_001013646.1, 1 a.a. - 174 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00201181-P01.10ug
Additional Details : Weight : 0.00010kg
Descripción
Sequence: MEGQRGAPRRSRGRPVSRGGAGHKAKRVTGGRGGRQGPSPAFHCALCQLQVNSETQLKQHMSSRRHKDRLAGKTPKPSSQHSKLQKHAALAVSILKSKLALQKQLTKTLAARFLPSPLPTAATAICALPGPLALRPAPTAATTLFPAPILGPALFRTPAGAVRPATGPIVLAPYEspecificaciones
NP_001013646.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ZNF385C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
201181 | |
LOC201181 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEGQRGAPRRSRGRPVSRGGAGHKAKRVTGGRGGRQGPSPAFHCALCQLQVNSETQLKQHMSSRRHKDRLAGKTPKPSSQHSKLQKHAALAVSILKSKLALQKQLTKTLAARFLPSPLPTAATAICALPGPLALRPAPTAATTLFPAPILGPALFRTPAGAVRPATGPIVLAPY | |
RUO | |
ZNF385C | |
Recombinant | |
wheat germ expression system |