missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LOC201181 Full-length ORF (NP_001013646.1, 1 a.a. - 174 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_001013646.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 201181 |
Molecular Weight (g/mol) | 44.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139273
|
Abnova™
H00201181-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16129273
|
Abnova™
H00201181-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sequence: MEGQRGAPRRSRGRPVSRGGAGHKAKRVTGGRGGRQGPSPAFHCALCQLQVNSETQLKQHMSSRRHKDRLAGKTPKPSSQHSKLQKHAALAVSILKSKLALQKQLTKTLAARFLPSPLPTAATAICALPGPLALRPAPTAATTLFPAPILGPALFRTPAGAVRPATGPIVLAPYSpecifications
NP_001013646.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MEGQRGAPRRSRGRPVSRGGAGHKAKRVTGGRGGRQGPSPAFHCALCQLQVNSETQLKQHMSSRRHKDRLAGKTPKPSSQHSKLQKHAALAVSILKSKLALQKQLTKTLAARFLPSPLPTAATAICALPGPLALRPAPTAATTLFPAPILGPALFRTPAGAVRPATGPIVLAPY | |
RUO | |
ZNF385C | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
201181 | |
LOC201181 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ZNF385C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |