missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human Laminin alpha 2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
1500.00 DKK - 3340.00 DKK
Tekniske data
| Antigen | Laminin alpha 2 |
|---|---|
| Klon | 1003531 |
| Fortynding | Immunohistochemistry 5-25 μg/mL, Immunocytochemistry 8-25 μg/mL |
| Applikationer | Immunohistochemistry, Immunocytochemistry |
| Klassifikation | Monoclonal |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
18751577
![]() |
R&D Systems
MAB2143-SP |
25 μg |
1500.00 DKK
25µg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
18722467
![]() |
R&D Systems
MAB2143-100 |
100 μg |
3340.00 DKK
100µg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
Laminin alpha 2 Monoclonal specifically detects Laminin alpha 2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Tekniske data
| Laminin alpha 2 | |
| Immunohistochemistry 5-25 μg/mL, Immunocytochemistry 8-25 μg/mL | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| LAMA2, Laminin M chain, laminin subunit alpha-2, laminin, alpha 2, Laminin-12 subunit alpha, Laminin-2 subunit alpha, LAMM, LAMMLaminin-4 subunit alpha, Merosin, Merosin heavy chain | |
| Chinese Hamster Ovary cell line, CHO-derived human Laminin alpha 2, Lys2145-Asn3122, Accession # P24043 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Laminin alpha 2 in direct ELISAs. |
| 1003531 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Human | |
| P24043 | |
| 3908 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel