missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human Laminin alpha 2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2143-100
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Laminin alpha 2 Monoclonal specifically detects Laminin alpha 2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Tekniske data
| Laminin alpha 2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| LAMA2, Laminin M chain, laminin subunit alpha-2, laminin, alpha 2, Laminin-12 subunit alpha, Laminin-2 subunit alpha, LAMM, LAMMLaminin-4 subunit alpha, Merosin, Merosin heavy chain | |
| Chinese Hamster Ovary cell line, CHO-derived human Laminin alpha 2, Lys2145-Asn3122, Accession # P24043 | |
| 100 μg | |
| 3908 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
| Immunohistochemistry, Immunocytochemistry | |
| 1003531 | |
| Immunohistochemistry 5-25 μg/mL, Immunocytochemistry 8-25 μg/mL | |
| P24043 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Laminin alpha 2 in direct ELISAs. | |
| Human | |
| Supernatant |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion