missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human IL-22 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
2315.00 DKK - 4185.00 DKK
Tekniske data
| Antigen | IL-22 |
|---|---|
| Klon | 142928 |
| Fortynding | Intracellular Staining by Flow Cytometry 10 uL/10^6 cells |
| Applikationer | Flow Cytometry |
| Klassifikation | Monoclonal |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
18734647
![]() |
R&D Systems
IC7821A-025 |
25 Tests |
2315.00 DKK
25 tests |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
18783187
![]() |
R&D Systems
IC7821A-100 |
100 Tests |
4185.00 DKK
100 tests |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Description
IL-22 Monoclonal specifically detects IL-22 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| IL-22 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
| IL22 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human IL-22 in direct ELISAs. In direct ELISAs, this antibody shows 100% cross-reactivity with recombinant mouse IL-22 and recombinant rat IL-22 and no cross-reactivity with recombinant human IL-10. |
| 142928 | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Human | |
| Q9GZX6 | |
| 50616 | |
| E. coli-derived recombinant human IL-22 Ala34-Ile179 Accession # Q9GZX6 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title