missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human IL-22 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC7821A-025
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IL-22 Monoclonal specifically detects IL-22 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Tekniske data
| IL-22 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 50616 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 142928 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Q9GZX6 | |
| IL22 | |
| E. coli-derived recombinant human IL-22 Ala34-Ile179 Accession # Q9GZX6 | |
| 25 Tests | |
| Primary | |
| Detects human IL-22 in direct ELISAs. In direct ELISAs, this antibody shows 100% cross-reactivity with recombinant mouse IL-22 and recombinant rat IL-22 and no cross-reactivity with recombinant human IL-10. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion