missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human Ig Kappa Light Chain Antibody, R&D Systems™

Mouse Monoclonal Antibody
1525.00 DKK - 3740.00 DKK
Tekniske data
| Antigen | Human Kappa Light Chain |
|---|---|
| Klon | 999916 |
| Fortynding | Western Blot 2 μg/mL, Simple Western 20 μg/mL, ELISA |
| Applikationer | Western Blot, ELISA |
| Klassifikation | Monoclonal |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
18779996
![]() |
R&D Systems
MAB10050-SP |
25 μg |
1525.00 DKK
25µg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
18781607
![]() |
R&D Systems
MAB10050-100 |
100 μg |
3740.00 DKK
100µg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
Human Kappa Light Chain Monoclonal specifically detects Human Kappa Light Chain in Human samples. It is validated for Western Blot, Simple Western, ELISA.Tekniske data
| Human Kappa Light Chain | |
| Western Blot 2 μg/mL, Simple Western 20 μg/mL, ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Human Ig Kappa Light Chain | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects Human Ig Kappa Light Chain in direct ELISAs and Western blots. |
| 999916 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel