missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human Ig Kappa Light Chain Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10050-100
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Human Kappa Light Chain Monoclonal specifically detects Human Kappa Light Chain in Human samples. It is validated for Western Blot, Simple Western, ELISA.
Tekniske data
| Human Kappa Light Chain | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Human Ig Kappa Light Chain | |
| 100 μg | |
| Detects Human Ig Kappa Light Chain in direct ELISAs and Western blots. | |
| Human | |
| Supernatant |
| Western Blot, ELISA | |
| 999916 | |
| Western Blot 2 μg/mL, Simple Western 20 μg/mL, ELISA | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion