Learn More
Abnova™ Human GLYCTK Full-length ORF (NP_660305.2, 1 a.a. - 523 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00132158-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This locus encodes a member of the glycerate kinase type-2 family. The encoded enzyme catalyzes the phosphorylation of (R)-glycerate and may be involved in serine degradation and fructose metabolism. Decreased activity of the encoded enzyme may be associated with the disease D-glyceric aciduria. Alternatively spliced transcript variants have been described. [provided by RefSeq]
Sequence: MAAALQVLPRLARAPLHPLLWRGSVARLASSMALAEQARQLFESAVGAVLPGPMLHRALSLDPGGRQLKVRDRNFQLRQNLYLVGFGKAVLGMAAAAEELLGQHLVQGVISVPKGIRAAMERAGKQEMLLKPHSRVQVFEGAEDNLPDRDALRAALAIQQLAEGLTADDLLLVLISGGGSALLPAPIPPVTLEEKQTLTRLLAARGATIQELNTIRKALSQLKGGGLAQAAYPAQVVSLILSDVVGDPVEVIASGPTVASSHNVQDCLHILNRYGLRAALPRSVKTVLSRADSDPHGPHTCGHVLNVIIGSNVLALAEAQRQAEALGYQAVVLSAAMQGDVKSMAQFYGLLAHVARTRLTPSMAGASVEEDAQLHELAAELQIPDLQLEEALETMAWGRGPVCLLAGGEPTVQLQGSGRGGRNQELALRVGAELRRWPLGPIDVLFLSGGTDGQDGPTEAAGAWVTPELASQAAAEGLDIATFLAHNDSHTFFCCLQGGAHLLHTGMTGTNVMDTHLLFLRPRSpecifications
NP_660305.2 | |
Liquid | |
132158 | |
GLYCTK (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAALQVLPRLARAPLHPLLWRGSVARLASSMALAEQARQLFESAVGAVLPGPMLHRALSLDPGGRQLKVRDRNFQLRQNLYLVGFGKAVLGMAAAAEELLGQHLVQGVISVPKGIRAAMERAGKQEMLLKPHSRVQVFEGAEDNLPDRDALRAALAIQQLAEGLTADDLLLVLISGGGSALLPAPIPPVTLEEKQTLTRLLAARGATIQELNTIRKALSQLKGGGLAQAAYPAQVVSLILSDVVGDPVEVIASGPTVASSHNVQDCLHILNRYGLRAALPRSVKTVLSRADSDPHGPHTCGHVLNVIIGSNVLALAEAQRQAEALGYQAVVLSAAMQGDVKSMAQFYGLLAHVARTRLTPSMAGASVEEDAQLHELAAELQIPDLQLEEALETMAWGRGPVCLLAGGEPTVQLQGSGRGGRNQELALRVGAELRRWPLGPIDVLFLSGGTDGQDGPTEAAGAWVTPELASQAAAEGLDIATFLAHNDSHTFFCCLQGGAHLLHTGMTGTNVMDTHLLFLRPR | |
RUO | |
GLYCTK | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
81.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GLYCTK1/HBEBP2/HBEBP4/HBeAgBP4A | |
GLYCTK | |
Yes | |
wheat germ expression system |