missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GLYCTK Full-length ORF (NP_660305.2, 1 a.a. - 523 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_660305.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 132158 |
Molecular Weight (g/mol) | 81.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16137963
|
Abnova™
H00132158-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16147963
|
Abnova™
H00132158-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This locus encodes a member of the glycerate kinase type-2 family. The encoded enzyme catalyzes the phosphorylation of (R)-glycerate and may be involved in serine degradation and fructose metabolism. Decreased activity of the encoded enzyme may be associated with the disease D-glyceric aciduria. Alternatively spliced transcript variants have been described. [provided by RefSeq]
Sequence: MAAALQVLPRLARAPLHPLLWRGSVARLASSMALAEQARQLFESAVGAVLPGPMLHRALSLDPGGRQLKVRDRNFQLRQNLYLVGFGKAVLGMAAAAEELLGQHLVQGVISVPKGIRAAMERAGKQEMLLKPHSRVQVFEGAEDNLPDRDALRAALAIQQLAEGLTADDLLLVLISGGGSALLPAPIPPVTLEEKQTLTRLLAARGATIQELNTIRKALSQLKGGGLAQAAYPAQVVSLILSDVVGDPVEVIASGPTVASSHNVQDCLHILNRYGLRAALPRSVKTVLSRADSDPHGPHTCGHVLNVIIGSNVLALAEAQRQAEALGYQAVVLSAAMQGDVKSMAQFYGLLAHVARTRLTPSMAGASVEEDAQLHELAAELQIPDLQLEEALETMAWGRGPVCLLAGGEPTVQLQGSGRGGRNQELALRVGAELRRWPLGPIDVLFLSGGTDGQDGPTEAAGAWVTPELASQAAAEGLDIATFLAHNDSHTFFCCLQGGAHLLHTGMTGTNVMDTHLLFLRPRSpecifications
NP_660305.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
81.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAAALQVLPRLARAPLHPLLWRGSVARLASSMALAEQARQLFESAVGAVLPGPMLHRALSLDPGGRQLKVRDRNFQLRQNLYLVGFGKAVLGMAAAAEELLGQHLVQGVISVPKGIRAAMERAGKQEMLLKPHSRVQVFEGAEDNLPDRDALRAALAIQQLAEGLTADDLLLVLISGGGSALLPAPIPPVTLEEKQTLTRLLAARGATIQELNTIRKALSQLKGGGLAQAAYPAQVVSLILSDVVGDPVEVIASGPTVASSHNVQDCLHILNRYGLRAALPRSVKTVLSRADSDPHGPHTCGHVLNVIIGSNVLALAEAQRQAEALGYQAVVLSAAMQGDVKSMAQFYGLLAHVARTRLTPSMAGASVEEDAQLHELAAELQIPDLQLEEALETMAWGRGPVCLLAGGEPTVQLQGSGRGGRNQELALRVGAELRRWPLGPIDVLFLSGGTDGQDGPTEAAGAWVTPELASQAAAEGLDIATFLAHNDSHTFFCCLQGGAHLLHTGMTGTNVMDTHLLFLRPR | |
RUO | |
GLYCTK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
132158 | |
GLYCTK (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GLYCTK1/HBEBP2/HBEBP4/HBeAgBP4A | |
GLYCTK | |
Recombinant | |
wheat germ expression system |