missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GIYD1 Full-length ORF (AAI48778.1, 1 a.a. - 161 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00548593-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16190784

  • 4040.00 DKK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A segmental duplication of the p arm of chromosome 16 created two identical copies of the GIY-YIG domain containing gene, this record represents the more centromeric copy. Exons of this gene overlap with exons of the phenol-preferring sulfotransferase (SULT1A3) gene. Two transcript variants that encode different protein isoforms have been identified through sequence analysis. [provided by RefSeq]

Sequence: MGPAGVAARPGRFFGVYLLYCLNPRYRGRVYVGFTVNTARRVQQHNGGRKKGGAWRTSGRGPWEMVLVVHGFPSSVAALRDEEGPLCCPHPGCLLRAHVICLAEEFLQEEPGQLLPLEGQCPCCEKSLLWGDLIWLCQMDTEKEVEDSELEEAHWTDLLET
Specifications

Specifications

AAI48778.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.66kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GIYD1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
548593
GIYD1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGPAGVAARPGRFFGVYLLYCLNPRYRGRVYVGFTVNTARRVQQHNGGRKKGGAWRTSGRGPWEMVLVVHGFPSSVAALRDEEGPLCCPHPGCLLRAHVICLAEEFLQEEPGQLLPLEGQCPCCEKSLLWGDLIWLCQMDTEKEVEDSELEEAHWTDLLET
RUO
GIYD1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers