missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GIYD1 Full-length ORF (AAI48778.1, 1 a.a. - 161 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | AAI48778.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 548593 |
Molecular Weight (g/mol) | 44.66kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16180784
|
Abnova™
H00548593-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16190784
|
Abnova™
H00548593-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
A segmental duplication of the p arm of chromosome 16 created two identical copies of the GIY-YIG domain containing gene, this record represents the more centromeric copy. Exons of this gene overlap with exons of the phenol-preferring sulfotransferase (SULT1A3) gene. Two transcript variants that encode different protein isoforms have been identified through sequence analysis. [provided by RefSeq]
Sequence: MGPAGVAARPGRFFGVYLLYCLNPRYRGRVYVGFTVNTARRVQQHNGGRKKGGAWRTSGRGPWEMVLVVHGFPSSVAALRDEEGPLCCPHPGCLLRAHVICLAEEFLQEEPGQLLPLEGQCPCCEKSLLWGDLIWLCQMDTEKEVEDSELEEAHWTDLLETSpecifications
AAI48778.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.66kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGPAGVAARPGRFFGVYLLYCLNPRYRGRVYVGFTVNTARRVQQHNGGRKKGGAWRTSGRGPWEMVLVVHGFPSSVAALRDEEGPLCCPHPGCLLRAHVICLAEEFLQEEPGQLLPLEGQCPCCEKSLLWGDLIWLCQMDTEKEVEDSELEEAHWTDLLET | |
RUO | |
GIYD1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
548593 | |
GIYD1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GIYD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |