missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FBXO15 Full-length ORF (NP_689889.1, 1 a.a. - 434 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00201456-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16139283

  • 4040.00 DKK / 25µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Members of the F-box protein family, such as FBXO15, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Sequence: MPSEILLKIFSYLDAVSLLCTGCVSRRFYHLANDNFIWIGIYSTAFSPARSNWKFNSVEKIAMSMSFLSVQDKEAGYWKKEYITKQIASVKAALADILKPVNPYTGLPVKTKEALRIFGLGWAIILKEKGGKEYIMEHVDLSINDTSVTVIWYGKKWPCLASLSTLDLCGMTPVFTDWYKTPTKHRLRWHSLIAKYNLSHLTISTMIGCDRLIRIFCLHPGLLVGVWKKEEELAFVMANLHFHHLVERSTLGSATIPYELPPHSPFLDDSPEYGLHGYQLHVDLHSGGVFYLCGTFRNLFTKRGNIENGHVKLIVIHLKNNREHLPLIGKVGLSWKTDIFDGCIKSCSMMDVTLLDEHGKPFWCFSSPVCLRSPATPSDSSSFLGQTYNVDYVDAEGRVHVELVWIRETEEYLIVNLVLYLSIAKINHWFGTEY
Specifications

Specifications

NP_689889.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
75.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FBX15/MGC39671
FBXO15
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
201456
FBXO15 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPSEILLKIFSYLDAVSLLCTGCVSRRFYHLANDNFIWIGIYSTAFSPARSNWKFNSVEKIAMSMSFLSVQDKEAGYWKKEYITKQIASVKAALADILKPVNPYTGLPVKTKEALRIFGLGWAIILKEKGGKEYIMEHVDLSINDTSVTVIWYGKKWPCLASLSTLDLCGMTPVFTDWYKTPTKHRLRWHSLIAKYNLSHLTISTMIGCDRLIRIFCLHPGLLVGVWKKEEELAFVMANLHFHHLVERSTLGSATIPYELPPHSPFLDDSPEYGLHGYQLHVDLHSGGVFYLCGTFRNLFTKRGNIENGHVKLIVIHLKNNREHLPLIGKVGLSWKTDIFDGCIKSCSMMDVTLLDEHGKPFWCFSSPVCLRSPATPSDSSSFLGQTYNVDYVDAEGRVHVELVWIRETEEYLIVNLVLYLSIAKINHWFGTEY
RUO
FBXO15
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers