missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FBXO15 Full-length ORF (NP_689889.1, 1 a.a. - 434 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_689889.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 201456 |
Molecular Weight (g/mol) | 75.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139283
|
Abnova™
H00201456-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16129283
|
Abnova™
H00201456-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Members of the F-box protein family, such as FBXO15, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]
Sequence: MPSEILLKIFSYLDAVSLLCTGCVSRRFYHLANDNFIWIGIYSTAFSPARSNWKFNSVEKIAMSMSFLSVQDKEAGYWKKEYITKQIASVKAALADILKPVNPYTGLPVKTKEALRIFGLGWAIILKEKGGKEYIMEHVDLSINDTSVTVIWYGKKWPCLASLSTLDLCGMTPVFTDWYKTPTKHRLRWHSLIAKYNLSHLTISTMIGCDRLIRIFCLHPGLLVGVWKKEEELAFVMANLHFHHLVERSTLGSATIPYELPPHSPFLDDSPEYGLHGYQLHVDLHSGGVFYLCGTFRNLFTKRGNIENGHVKLIVIHLKNNREHLPLIGKVGLSWKTDIFDGCIKSCSMMDVTLLDEHGKPFWCFSSPVCLRSPATPSDSSSFLGQTYNVDYVDAEGRVHVELVWIRETEEYLIVNLVLYLSIAKINHWFGTEYSpecifications
NP_689889.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
75.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MPSEILLKIFSYLDAVSLLCTGCVSRRFYHLANDNFIWIGIYSTAFSPARSNWKFNSVEKIAMSMSFLSVQDKEAGYWKKEYITKQIASVKAALADILKPVNPYTGLPVKTKEALRIFGLGWAIILKEKGGKEYIMEHVDLSINDTSVTVIWYGKKWPCLASLSTLDLCGMTPVFTDWYKTPTKHRLRWHSLIAKYNLSHLTISTMIGCDRLIRIFCLHPGLLVGVWKKEEELAFVMANLHFHHLVERSTLGSATIPYELPPHSPFLDDSPEYGLHGYQLHVDLHSGGVFYLCGTFRNLFTKRGNIENGHVKLIVIHLKNNREHLPLIGKVGLSWKTDIFDGCIKSCSMMDVTLLDEHGKPFWCFSSPVCLRSPATPSDSSSFLGQTYNVDYVDAEGRVHVELVWIRETEEYLIVNLVLYLSIAKINHWFGTEY | |
RUO | |
FBXO15 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
201456 | |
FBXO15 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBX15/MGC39671 | |
FBXO15 | |
Recombinant | |
wheat germ expression system |