Learn More
Abnova™ Human EDIL3 Partial ORF (NP_005702, 101 a.a. - 199 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010085-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is an integrin ligand. It plays an important role in mediating angiogenesis and may be important in vessel wall remodeling and development. It also influences endothelial cell behavior. [provided by RefSeq]
Sequence: IGYVCKCPRGFNGIHCQHNINECEVEPCKNGGICTDLVANYSCECPGEFMGRNCQYKCSGPLGIEGGIISNQQITASSTHRALFGLQKWYPYYARLNKKSpecifications
NP_005702 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IGYVCKCPRGFNGIHCQHNINECEVEPCKNGGICTDLVANYSCECPGEFMGRNCQYKCSGPLGIEGGIISNQQITASSTHRALFGLQKWYPYYARLNKK | |
RUO | |
EDIL3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10085 | |
EDIL3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DEL1/MGC26287 | |
EDIL3 | |
Recombinant | |
wheat germ expression system |