missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EDIL3 Partial ORF (NP_005702, 101 a.a. - 199 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

2665.00 DKK - 4040.00 DKK

Specifications

Accession Number NP_005702
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10085
Molecular Weight (g/mol) 36.63kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16131826
View Documents Promotion Details
Abnova™
H00010085-Q01.25UG
25 ug
4040.00 DKK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16121826
View Documents Promotion Details
Abnova™
H00010085-Q01.10UG
10 ug
2665.00 DKK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene is an integrin ligand. It plays an important role in mediating angiogenesis and may be important in vessel wall remodeling and development. It also influences endothelial cell behavior. [provided by RefSeq]

Sequence: IGYVCKCPRGFNGIHCQHNINECEVEPCKNGGICTDLVANYSCECPGEFMGRNCQYKCSGPLGIEGGIISNQQITASSTHRALFGLQKWYPYYARLNKK
Specifications

Specifications

NP_005702
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DEL1/MGC26287
EDIL3
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
10085
EDIL3 (Human) Recombinant Protein (Q01)
IGYVCKCPRGFNGIHCQHNINECEVEPCKNGGICTDLVANYSCECPGEFMGRNCQYKCSGPLGIEGGIISNQQITASSTHRALFGLQKWYPYYARLNKK
RUO
EDIL3
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications