missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DISP1 Full-length ORF (AAH07734.1, 1 a.a. - 542 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084976-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136243

  • 2735.00 DKK / 10µg
Estimated Shipment: 24-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The pattern of cellular proliferation and differentiation that leads to normal development of embryonic structures often depends upon the localized production of secreted protein signals. Cells surrounding the source of a particular signal respond in a graded manner according to the effective concentration of the signal, and this response produces the pattern of cell types constituting the mature structure. A novel segment-polarity gene known as dispatched has been identified in Drosophila and its protein product is required for normal Hedgehog (Hh) signaling. This gene is one of two human homologs of Drosophila dispatched and, based on sequence identity to its mouse counterpart, the encoded protein may play an essential role in Hh patterning activities in the early embryo. [provided by RefSeq]

Sequence: MAMSNGNNDFVVLSNSSIATSAANPSPLTPCDGDHAAQQLTPKEATRTKVSPNGCLQLNGTVKSSFLPLDNQRMPQMLPQCCHPCPYHHPLTSHSSHQECHPEAGPAAPSALASCCMQPHSEYSASLCPNHSPVYQTTCCLQPSPSFCLHHPWPDHFQHQPVQQHIANIRPSRPFKLPKSYAALIADWPVVVLGMCTMFIVVCALVGVLVPELPDFSDPLLGFEPRGTAIGQRLVTWNNMVKNTGYKATLANYPFKYADEQAKSHRDDRWSDDHYEREKREVDWNFHKDSFFCDVPSDRYSRVVFTSSGGETLWNLPAIKSMCNVDNSRIRSHPQFGDLCQRTTAASCCPSWTLGNYIAILNNRSSCQKIVERDVSHTLKLLRTCAKHYQNGTLGPDCWDMAARRKDQLKCTNVPRKCTKYNAVYQILHYLVDKDFMTPKTADYATPALKYSMLFSPTEKGESMMNIYLDNFENWNSSDGVTTITGIEFGIKHSLFQDYLLMDTVYPAIAIVIVLLVMCVYTKSMFITLMTMFAIISSLIVS
Specifications

Specifications

AAH07734.1
Liquid
84976
DISP1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAMSNGNNDFVVLSNSSIATSAANPSPLTPCDGDHAAQQLTPKEATRTKVSPNGCLQLNGTVKSSFLPLDNQRMPQMLPQCCHPCPYHHPLTSHSSHQECHPEAGPAAPSALASCCMQPHSEYSASLCPNHSPVYQTTCCLQPSPSFCLHHPWPDHFQHQPVQQHIANIRPSRPFKLPKSYAALIADWPVVVLGMCTMFIVVCALVGVLVPELPDFSDPLLGFEPRGTAIGQRLVTWNNMVKNTGYKATLANYPFKYADEQAKSHRDDRWSDDHYEREKREVDWNFHKDSFFCDVPSDRYSRVVFTSSGGETLWNLPAIKSMCNVDNSRIRSHPQFGDLCQRTTAASCCPSWTLGNYIAILNNRSSCQKIVERDVSHTLKLLRTCAKHYQNGTLGPDCWDMAARRKDQLKCTNVPRKCTKYNAVYQILHYLVDKDFMTPKTADYATPALKYSMLFSPTEKGESMMNIYLDNFENWNSSDGVTTITGIEFGIKHSLFQDYLLMDTVYPAIAIVIVLLVMCVYTKSMFITLMTMFAIISSLIVS
RUO
DISP1
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
87.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DISPA/DKFZp434I0428/FLJ43740/MGC104180/MGC13130/MGC16796
DISP1
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers