Learn More
Abnova™ Human DISP1 Full-length ORF (AAH07734.1, 1 a.a. - 542 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | AAH07734.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 84976 |
Molecular Weight (g/mol) | 87.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16146243
|
Abnova™
H00084976-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16136243
|
Abnova™
H00084976-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The pattern of cellular proliferation and differentiation that leads to normal development of embryonic structures often depends upon the localized production of secreted protein signals. Cells surrounding the source of a particular signal respond in a graded manner according to the effective concentration of the signal, and this response produces the pattern of cell types constituting the mature structure. A novel segment-polarity gene known as dispatched has been identified in Drosophila and its protein product is required for normal Hedgehog (Hh) signaling. This gene is one of two human homologs of Drosophila dispatched and, based on sequence identity to its mouse counterpart, the encoded protein may play an essential role in Hh patterning activities in the early embryo. [provided by RefSeq]
Sequence: MAMSNGNNDFVVLSNSSIATSAANPSPLTPCDGDHAAQQLTPKEATRTKVSPNGCLQLNGTVKSSFLPLDNQRMPQMLPQCCHPCPYHHPLTSHSSHQECHPEAGPAAPSALASCCMQPHSEYSASLCPNHSPVYQTTCCLQPSPSFCLHHPWPDHFQHQPVQQHIANIRPSRPFKLPKSYAALIADWPVVVLGMCTMFIVVCALVGVLVPELPDFSDPLLGFEPRGTAIGQRLVTWNNMVKNTGYKATLANYPFKYADEQAKSHRDDRWSDDHYEREKREVDWNFHKDSFFCDVPSDRYSRVVFTSSGGETLWNLPAIKSMCNVDNSRIRSHPQFGDLCQRTTAASCCPSWTLGNYIAILNNRSSCQKIVERDVSHTLKLLRTCAKHYQNGTLGPDCWDMAARRKDQLKCTNVPRKCTKYNAVYQILHYLVDKDFMTPKTADYATPALKYSMLFSPTEKGESMMNIYLDNFENWNSSDGVTTITGIEFGIKHSLFQDYLLMDTVYPAIAIVIVLLVMCVYTKSMFITLMTMFAIISSLIVSSpecifications
AAH07734.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
87.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAMSNGNNDFVVLSNSSIATSAANPSPLTPCDGDHAAQQLTPKEATRTKVSPNGCLQLNGTVKSSFLPLDNQRMPQMLPQCCHPCPYHHPLTSHSSHQECHPEAGPAAPSALASCCMQPHSEYSASLCPNHSPVYQTTCCLQPSPSFCLHHPWPDHFQHQPVQQHIANIRPSRPFKLPKSYAALIADWPVVVLGMCTMFIVVCALVGVLVPELPDFSDPLLGFEPRGTAIGQRLVTWNNMVKNTGYKATLANYPFKYADEQAKSHRDDRWSDDHYEREKREVDWNFHKDSFFCDVPSDRYSRVVFTSSGGETLWNLPAIKSMCNVDNSRIRSHPQFGDLCQRTTAASCCPSWTLGNYIAILNNRSSCQKIVERDVSHTLKLLRTCAKHYQNGTLGPDCWDMAARRKDQLKCTNVPRKCTKYNAVYQILHYLVDKDFMTPKTADYATPALKYSMLFSPTEKGESMMNIYLDNFENWNSSDGVTTITGIEFGIKHSLFQDYLLMDTVYPAIAIVIVLLVMCVYTKSMFITLMTMFAIISSLIVS | |
RUO | |
DISP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
84976 | |
DISP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DISPA/DKFZp434I0428/FLJ43740/MGC104180/MGC13130/MGC16796 | |
DISP1 | |
Recombinant | |
wheat germ expression system |