Få mere at vide
Medicom™ Hopen™ Single Use Hygiene Gown
Medicom™ Hopen™ Single Use Hygiene Gown without pockets, is microporous and waterproof
Brand: Medicom™ 40.550/04
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
- Reduces contamination of the operator, but also sensitive products and processes in a protected atmosphere
- Protects against radioactive particles and infectious agents
- Microporous, Anti-Static Hygiene Gown is comfortable and light
- Polypropylene gown lined with laminated polyethylene for a waterproof, hydrophobic seal
- Available in mulitple sizes, and closure types
Tekniske data
| X-Large | |
| Polypropylene | |
| Press studs | |
| Yes | |
| Radioactive Particles, Infectious Agents | |
| Microporous, Waterproof | |
| Manufactured under an ISO 9001 Certified Quality System | |
| Category 1 Personal Protective Equipment |
| Unisex | |
| White | |
| Elastic | |
| Non-sterile | |
| None | |
| Sewn | |
| EN 1149-5 | |
| Individual, Bag |