Learn More
Honeywell™ Supervizor Face Shields
Robust, heavy-duty, helmet-mounted face shield range with hearing protection compatibility. Sperian™ Honeywell™ Supervizor Face Shields have a wide visor selection that can be tailored to the specific hazard/application.
Brand: Honeywell™ 1002297
Additional Details : Weight : 0.21000kg
Description
- Recommended for use in the chemical and forestry industries, among others
- Protects against hazards such as abrasion, airborne particles, biohazards, chemicals, gases, vapors, and smoke
- Maximum protection, allowing full unrestricted head movement
- Creased visor eliminates gaps and visor profile follows contours of the face
Specifications
Browguard with Ratchet Headband | |
Supervizor Faceshield | |
1 Ea. | |
Polycarbonate |