Learn More
Brady™ High Density Thermal Transfer Printer Ribbon
High Density Thermal Transfer Printer Ribbon with excellent solvent resistance, high heat resistance, and super scratch resistance.
Brand: Brady™ 114553
Additional Details : Weight : 0.11790kg
Description
- High density ribbon with excellent solvent resistance, high heat resistance, and super scratch resistance.
Specifications
Printer Ribbon | |
BMP71 Label Printer | |
1.97 in. | |
50 mm | |
Green 6900 Series Thermal Transfer Printer Ribbon for BMP71 Printer |
Green | |
50 yd. | |
45.72 m | |
46 m x 50.8 mm | |
1 Ea. |